Stampa…

Tetraethylammonium fluoride 98%

Fornitore: Apollo Scientific

PC903855-100G PC903855-25G
APOSPC903855-100GEA 270 EUR
APOSPC903855-100G APOSPC903855-25G
Tetraethylammonium fluoride 98%
Tetraethylammonium fluoride
Formula: C₈H₂₀FN
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011826
CAS Number: 665-46-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR