Stampa…

Methyl-2-amino-4,5-difluorobenzoate 98%

Fornitore: Apollo Scientific

Synonyms: Methyl 2-amino-4,5-difluorobenzoate

PC902903-100G PC902903-500G
APOSPC902903-100GEA 170 EUR
APOSPC902903-100G APOSPC902903-500G
Methyl-2-amino-4,5-difluorobenzoate 98%
Methyl-2-amino-4,5-difluorobenzoate
Formula: C₈H₇F₂NO₂
Peso molecolare: 187,15 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09264511
CAS Number: 207346-42-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR