Stampa…

3-(4-Fluorophenyl)acrylaldehyde 95%

Fornitore: Apollo Scientific

Synonyms: 3-(4-Fluorophenyl)-2-propenal, 4-Fluorocinnamaldehyde, 3-(4-Fluorophenyl)acrylaldehyde

PC902652-100G PC902652-500G
APOSPC902652-100GEA 168 EUR
APOSPC902652-100G APOSPC902652-500G
3-(4-Fluorophenyl)acrylaldehyde 95%
3-(4-Fluorophenyl)acrylaldehyde
Formula: C₉H₇FO
Peso molecolare: 150,15 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03425895
CAS Number: 24654-55-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR