Stampa…

Ethyl-3-amino-4-fluorobenzoate 95%

Fornitore: Apollo Scientific

Synonyms: Ethyl 3-amino-4-fluorobenzoate

PC902645-10G PC902645-25G PC902645-5G
APOSPC902645-10GEA 183 EUR
APOSPC902645-10G APOSPC902645-25G APOSPC902645-5G
Ethyl-3-amino-4-fluorobenzoate 95%
Ethyl-3-amino-4-fluorobenzoate
Formula: C₉H₁₀FNO₂
Peso molecolare: 183,1796 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03840487
CAS Number: 455-75-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR