Stampa…

6-Fluoro-1-methylbenzimidazole 95%

Fornitore: Apollo Scientific

Synonyms: 6-Fluoro-1-methyl-1H-benzo[d]imidazole

PC902247-1G PC902247-5G
APOSPC902247-1GEA 219 EUR
APOSPC902247-1G APOSPC902247-5G
6-Fluoro-1-methylbenzimidazole 95%
6-Fluoro-1-methylbenzimidazole
Formula: C₈H₇FN₂
Peso molecolare: 150,15 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD12546503
CAS Number: 1187385-86-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR