8-(Trifluoromethyl)quinoline 98%
Fornitore: Apollo Scientific
PC901829-10G
PC901829-1G
PC901829-5G
APOSPC901829-10GEA
887
EUR
APOSPC901829-10G
APOSPC901829-1G
APOSPC901829-5G
8-(Trifluoromethyl)quinoline 98%
8-(Trifluoromethyl)quinoline
Formula:
C₁₀H₆F₃N Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09744668 CAS Number: 317-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...