Ethyl-1-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate 97%
Fornitore: Apollo Scientific
PC8473-1G
PC8473-500MG
PC8473-5G
APOSPC8473-1GEA
533
EUR
APOSPC8473-1G
APOSPC8473-500MG
APOSPC8473-5G
Ethyl-1-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate 97%
Ethyl-1-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Formula:
C₁₆H₁₁ClF₃N₃O₂S |
MDL Number:
MFCD00665229 CAS Number: 159885-63-9 |