Diphenyliodonium hexafluorophosphate 98%
Fornitore: Apollo Scientific
|
Pericolo
|
PC8214-50G
PC8214-10G
PC8214-250G
APOSPC8214-50GEA
357
EUR
APOSPC8214-50G
APOSPC8214-10G
APOSPC8214-250G
Diphenyliodonium hexafluorophosphate 98%
Diphenyliodonium hexafluorophosphate
Formula:
C₁₂H₁₀F₆IP Peso molecolare: 426,08 g/mol Punto di fusione: 142 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00061398 CAS Number: 58109-40-3 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...