2-Bromo-N-(4-fluorobenzyl)benzenesulphonamide
Fornitore: Apollo Scientific
PC8079-1G
PC8079-5G
APOSPC8079-1GEA
132
EUR
APOSPC8079-1G
APOSPC8079-5G
2-Bromo-N-(4-fluorobenzyl)benzenesulphonamide
2-Bromo-N-(4-fluorobenzyl)benzenesulphonamide
Formula:
C₁₃H₁₁BrFNO₂S Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD02556038 CAS Number: 321705-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...