Stampa…

Ultramark® 1621

Fornitore: Apollo Scientific
Attenzione

Synonyms: Massmark 1621

PC7950-100MG PC7950-1G PC7950-250MG
APOSPC7950-100MGEA 0 EUR
APOSPC7950-100MG APOSPC7950-1G APOSPC7950-250MG
Ultramark® 1621
Ultramark® 1621

Fluorinated phosphazene for MS, especially suitable for +ve and -ve FAB high resolution MS (Jiang & Moini, ASMS Poster 1992), showing peaks upto 2120 mass units. R= -CH2(CF2CF2)xH, x=1,2 or 3; Fluorinated mass marker up to mass 2120

Formula: C₆H₁₈N₃P₃
Peso molecolare: 231,189 g/mol
Punto di ebollizione: 160…220 °C/0,01mmHg
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00242454
CAS Number: 105809-15-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR