Stampa…

2-Bromo-5-(trifluoromethyl)phenylboronic acid 96%

Fornitore: Apollo Scientific
Attenzione

Synonyms: (2-Bromo-5-(trifluoromethyl)phenyl)boronic acid, 2-Bromo-5-trifluoromethylphenylboronic acid

PC7451-1G PC7451-5G
APOSPC7451-1GEA 91.6 EUR
APOSPC7451-1G APOSPC7451-5G
2-Bromo-5-(trifluoromethyl)phenylboronic acid 96%
2-Bromo-5-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BBrF₃O₂
Peso molecolare: 268,82 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD09258742
CAS Number: 957034-38-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR