Stampa…

Sodium trifluoromethanesulphonate 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Trifluoromethanesulphonic acid sodium salt

PC6572-25G PC6572-5G PC6572-100G
APOSPC6572-25GEA 65.5 EUR
APOSPC6572-25G APOSPC6572-5G APOSPC6572-100G
Sodium trifluoromethanesulphonate 98%
Sodium trifluoromethanesulphonate
Formula: CF₃NaO₃S
Peso molecolare: 172,06 g/mol
Punto di fusione: 253…255 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00061607
CAS Number: 2926-30-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR