Stampa…

Phenyl trifluoromethyl sulphide 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Phenyl(trifluoromethyl)sulphane, alpha,alpha,alpha-Trifluorothioanisole, (Trifluoromethylthio)benzene

PC6229-25G PC6229-5G
APOSPC6229-25GEA 246 EUR
APOSPC6229-25G APOSPC6229-5G
Phenyl trifluoromethyl sulphide 98%
Phenyl trifluoromethyl sulphide
Formula: C₇H₅F₃S
Peso molecolare: 178,17 g/mol
Punto di ebollizione: 140…142 °C
Densità: 1,249 g/cm³ (25 °C)
Flash Pt: 37 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00040839
CAS Number: 456-56-4
EINECS: 207-270-0
UN: 1993
ADR: 3,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR