Stampa…

4-Chloro-3-fluoropyridine hydrochloride

Fornitore: Apollo Scientific

PC5997-1G PC5997-5G
APOSPC5997-1GEA 71.3 EUR
APOSPC5997-1G APOSPC5997-5G
4-Chloro-3-fluoropyridine hydrochloride
4-Chloro-3-fluoropyridine hydrochloride
Formula: C₅H₄Cl₂FN
Flash Pt: >110 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03092936
CAS Number: 119229-74-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR