1-(4-Fluoro-2-nitrophenyl)-3-methylpiperidine
Fornitore: Apollo Scientific
PC5315-1G
PC5315-5G
APOSPC5315-1GEA
114
EUR
APOSPC5315-1G
APOSPC5315-5G
1-(4-Fluoro-2-nitrophenyl)-3-methylpiperidine
1-(4-Fluoro-2-nitrophenyl)-3-methylpiperidine
Formula:
C₁₂H₁₅FN₂O₂ Punto di fusione: 31…32 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD08692445 CAS Number: 942474-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...