Stampa…

2,6-Dichloro-3-(difluoromethyl)pyridine 95%

Fornitore: Apollo Scientific

PC520760-10G PC520760-1G PC520760-5G
APOSPC520760-10GEA 1470 EUR
APOSPC520760-10G APOSPC520760-1G APOSPC520760-5G
2,6-Dichloro-3-(difluoromethyl)pyridine 95%
2,6-Dichloro-3-(difluoromethyl)pyridine
Formula: C₆H₃Cl₂F₂N
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD22199294
CAS Number: 1374659-26-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR