Stampa…

3-Chloro-4-(difluoromethyl)pyridine 97%

Fornitore: Apollo Scientific

PC520755-10G PC520755-1G PC520755-5G
APOSPC520755-10GEA 1630 EUR
APOSPC520755-10G APOSPC520755-1G APOSPC520755-5G
3-Chloro-4-(difluoromethyl)pyridine 97%
3-Chloro-4-(difluoromethyl)pyridine
Formula: C₆H₄ClF₂N
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD22124921
CAS Number: 1374659-44-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR