(1-Benzyl-4,4-difluoropiperidin-3-yl)methanol 95%
Fornitore: Apollo Scientific
Formula:
C₁₃H₁₇F₂NO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD18909828 CAS Number: 1303973-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...