Stampa…

1,3-Dichloro-2-(3,3,3-trifluoropropyl)benzene 95%

Fornitore: Apollo Scientific

PC520651-1G PC520651-5G
APOSPC520651-1GEA 193 EUR
APOSPC520651-1G APOSPC520651-5G
1,3-Dichloro-2-(3,3,3-trifluoropropyl)benzene 95%
1,3-Dichloro-2-(3,3,3-trifluoropropyl)benzene
Formula: C₉H₇Cl₂F₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11226648
CAS Number: 1099597-55-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR