Stampa…

1-Bromo-4-(2,2,2-trifluoroethyl)benzene 97%

Fornitore: Apollo Scientific

PC520615-25G PC520615-5G
APOSPC520615-25GEA 1050 EUR
APOSPC520615-25G APOSPC520615-5G
1-Bromo-4-(2,2,2-trifluoroethyl)benzene 97%
1-Bromo-4-(2,2,2-trifluoroethyl)benzene
Formula: C₈H₆BrF₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD10697865
CAS Number: 155820-88-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR