Stampa…

1-Methyl-4-(pentafluoroethyl)benzene 95%

Fornitore: Apollo Scientific

PC520584-1G PC520584-5G
APOSPC520584-1GEA 118 EUR
APOSPC520584-1G APOSPC520584-5G
1-Methyl-4-(pentafluoroethyl)benzene 95%
1-Methyl-4-(pentafluoroethyl)benzene
Formula: C₉H₇F₅
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09909908
CAS Number: 117081-46-6

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR