2',6'-Difluoro-4'-methoxyacetophenone 97%
Fornitore: Apollo Scientific
PC520530-1G
PC520530-5G
APOSPC520530-1GEA
152
EUR
APOSPC520530-1G
APOSPC520530-5G
2',6'-Difluoro-4'-methoxyacetophenone 97%
2',6'-Difluoro-4'-methoxyacetophenone
Formula:
C₉H₈F₂O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04115921 CAS Number: 886498-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...