1-(2-Fluorophenyl)cyclopentanecarboxylic acid 98%
Fornitore: Apollo Scientific
Formula:
C₁₂H₁₃FO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00800622 CAS Number: 214262-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...