3,6-Dichloro-2-(trifluoromethyl)pyridine 97%
Fornitore: Apollo Scientific
PC520506-10G
PC520506-1G
PC520506-5G
APOSPC520506-10GEA
844
EUR
APOSPC520506-10G
APOSPC520506-1G
APOSPC520506-5G
3,6-Dichloro-2-(trifluoromethyl)pyridine 97%
3,6-Dichloro-2-(trifluoromethyl)pyridine
Formula:
C₆H₂Cl₂F₃N Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00525616 CAS Number: 89719-91-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...