Stampa…

4-Bromo-3-fluorobenzyl alcohol

Fornitore: Apollo Scientific

Synonyms: (4-Bromo-3-fluorophenyl)methanol

PC501319-1G PC501319-5G
APOSPC501319-1GEA 53.9 EUR
APOSPC501319-1G APOSPC501319-5G
4-Bromo-3-fluorobenzyl alcohol
4-Bromo-3-fluorobenzyl alcohol
Formula: C₇H₆BrFO
Peso molecolare: 205,026 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08236860
CAS Number: 222978-01-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR