4-Bromo-2,5-difluorobenzonitrile
Fornitore: Apollo Scientific
|
Attenzione
|
PC49639-1G
PC49639-25G
PC49639-5G
APOSPC49639-1GEA
26.3
EUR
APOSPC49639-1G
APOSPC49639-25G
APOSPC49639-5G
4-Bromo-2,5-difluorobenzonitrile
4-Bromo-2,5-difluorobenzonitrile
Formula:
C₇H₂BrF₂N Peso molecolare: 218,0000 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11847049 CAS Number: 133541-45-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...