Stampa…

6-Chloro-5-fluoropyrimidin-4-amine

Fornitore: Apollo Scientific

Synonyms: 4-Amino-6-chloro-5-fluoropyrimidine

PC49628-1G PC49628-5G
APOSPC49628-1GEA 56.8 EUR
APOSPC49628-1G APOSPC49628-5G
6-Chloro-5-fluoropyrimidin-4-amine
6-Chloro-5-fluoropyrimidin-4-amine
Formula: C₄H₃ClFN₃
Peso molecolare: 147,5400 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD14706712
CAS Number: 851984-15-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR