4-Bromo-2-(trifluoromethyl)quinoline 96%
Fornitore: Apollo Scientific
|
Attenzione
|
PC49117-1G
PC49117-250MG
PC49117-5G
APOSPC49117-1GEA
78.6
EUR
APOSPC49117-1G
APOSPC49117-250MG
APOSPC49117-5G
4-Bromo-2-(trifluoromethyl)quinoline 96%
4-Bromo-2-(trifluoromethyl)quinoline
Formula:
C₁₀H₅BrF₃N Peso molecolare: 276,0526 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00277488 CAS Number: 18706-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...