2,3,5,6-Tetrafluoroisonicotinic acid 98%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
Perfluoroisonicotinic acid
PC4551-1G
PC4551-5G
APOSPC4551-1GEA
132
EUR
APOSPC4551-1G
APOSPC4551-5G
2,3,5,6-Tetrafluoroisonicotinic acid 98%
2,3,5,6-Tetrafluoroisonicotinic acid
Formula:
C₆HF₄NO₂ Peso molecolare: 195,07 g/mol Punto di fusione: 98…100 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD01320762 CAS Number: 2875-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...