Stampa…

2-Trifluoromethyl-1H-benzoimidazole-5-carboxylic acid

Fornitore: Apollo Scientific

PC450057-1G PC450057-5G
APOSPC450057-1GEA 109 EUR
APOSPC450057-1G APOSPC450057-5G
2-Trifluoromethyl-1H-benzoimidazole-5-carboxylic acid
2-Trifluoromethyl-1H-benzoimidazole-5-carboxylic acid
Formula: C₉H₅F₃N₂O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01309785
CAS Number: 82791-93-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR