Stampa…

4-Chloro-2-methyl-6-(trifluoromethyl)pyrimidine 95+%

Fornitore: Apollo Scientific

PC446096-1G PC446096-500MG PC446096-5G
APOSPC446096-1GEA 161 EUR
APOSPC446096-1G APOSPC446096-500MG APOSPC446096-5G
4-Chloro-2-methyl-6-(trifluoromethyl)pyrimidine 95+%
4-Chloro-2-methyl-6-(trifluoromethyl)pyrimidine
Formula: C₆H₄ClF₃N₂
Peso molecolare: 196,5576 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03265399
CAS Number: 5993-98-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR