Stampa…

2-(4-Fluorophenyl)thiazole-4-carboxylic acid

Fornitore: Apollo Scientific

PC446054-1G PC446054-5G
APOSPC446054-1GEA 183 EUR
APOSPC446054-1G APOSPC446054-5G
2-(4-Fluorophenyl)thiazole-4-carboxylic acid
2-(4-Fluorophenyl)thiazole-4-carboxylic acid
Formula: C₁₀H₆FNO₂S
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD07339052
CAS Number: 863668-07-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR