Stampa…

N'-[2-(4-Methoxyphenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl

Fornitore: Apollo Scientific

PC446010-1G PC446010-500MG
APOSPC446010-1GEA 183 EUR
APOSPC446010-1G APOSPC446010-500MG
N'-[2-(4-Methoxyphenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl
N'-[2-(4-Methoxyphenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl
Formula: C₁₆H₂₂F₃N₃O₃
Peso molecolare: 361,37 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD10568255
CAS Number: 1053657-61-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR