Stampa…

2-Chloro-6-fluorobenzothiazole

Fornitore: Apollo Scientific

Synonyms: 2-Chloro-6-fluorobenzo[d]thiazole

PC430516-1G PC430516-5G
APOSPC430516-1GEA 19 EUR
APOSPC430516-1G APOSPC430516-5G
2-Chloro-6-fluorobenzothiazole
2-Chloro-6-fluorobenzothiazole
Formula: C₇H₃ClFNS
Peso molecolare: 187,62 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04448822
CAS Number: 399-74-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR