Stampa…

5-Fluoro-1H-1,2,3-benzotriazole 95%

Fornitore: Apollo Scientific

Synonyms: 5-Fluoro-1H-benzo[d][1,2,3]triazole

PC421222-1G PC421222-250MG PC421222-5G
APOSPC421222-1GEA 314 EUR
APOSPC421222-1G APOSPC421222-250MG APOSPC421222-5G
5-Fluoro-1H-1,2,3-benzotriazole 95%
5-Fluoro-1H-1,2,3-benzotriazole
Formula: C₆H₄FN₃
Peso molecolare: 137,12 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00760403
CAS Number: 18225-90-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR