Stampa…

α-Fluorophenylacetic acid 97%

Fornitore: Apollo Scientific
Pericolo

Synonyms: 2-Fluoro-2-phenylacetic acid, alpha-Fluorophenylacetic acid

PC4119-1G PC4119-5G
APOSPC4119-1GEA 161 EUR
APOSPC4119-1G APOSPC4119-5G
α-Fluorophenylacetic acid 97%
α-Fluorophenylacetic acid
Formula: C₈H₇FO₂
Peso molecolare: 154.14 g/mol
Punto di fusione: 78…85 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004221
CAS Number: 1578-63-8
EINECS: 216-420-4
UN: 2811
ADR: 6.1 III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR