(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate
Fornitore: Apollo Scientific
Synonyms:
1-tert-butyl 2-methyl (2S)-4,4-difluoropyrrolidine-1,2-dicarboxylate, (S)-1-tert-Butyl 2-methyl 4,4-difluoropyrrolidine-1,2-dicarboxylate
PC409002-1G
PC409002-250MG
APOSPC409002-1GEA
90.2
EUR
APOSPC409002-1G
APOSPC409002-250MG
(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate
(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate
Formula:
C₁₁H₁₇F₂NO₄ Peso molecolare: 265,26 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03094918 CAS Number: 203866-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...