Stampa…

Dichlorofluoroacetic acid

Fornitore: Apollo Scientific
Pericolo

PC408090-25G PC408090-5G
APOSPC408090-25GEA 651 EUR
APOSPC408090-25G APOSPC408090-5G
Dichlorofluoroacetic acid
Dichlorofluoroacetic acid
Formula: C₂HCl₂FO₂
Temperatura di stoccaggio: Ambiente
CAS Number: 354-19-8
EINECS: 206-547-3
UN: 3265
ADR: 8,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR