Stampa…

4-Fluoro-N-(4-methoxybenzyl)benzamide

Fornitore: Apollo Scientific

PC3661-1G PC3661-5G
APOSPC3661-1GEA 72.7 EUR
APOSPC3661-1G APOSPC3661-5G
4-Fluoro-N-(4-methoxybenzyl)benzamide
4-Fluoro-N-(4-methoxybenzyl)benzamide
Formula: C₁₅H₁₄FNO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01359589
CAS Number: 346725-04-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR