Stampa…

2-Chloro-5-fluoro-4-pyridineboronic acid

Fornitore: Apollo Scientific
Attenzione

Synonyms: 2-Chloro-5-fluoropyridine-4-boronic Acid

PC3618-250MG PC3618-1G PC3618-5G
APOSPC3618-250MGEA 81.5 EUR
APOSPC3618-250MG APOSPC3618-1G APOSPC3618-5G
2-Chloro-5-fluoro-4-pyridineboronic acid
2-Chloro-5-fluoro-4-pyridineboronic acid
Formula: C₅H₄BClFNO₂
Peso molecolare: 175,35 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD04972404
CAS Number: 951677-47-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR