Stampa…

2-Fluoro-3-methyl-5-pyridineboronic acid

Fornitore: Apollo Scientific

Synonyms: 6-Fluoro-5-methyl-3-pyridylboronic acid, 2-Fluoro-3-methylpyridine-5-boronic acid, (6-Fluoro-5-methylpyridin-3-yl)boronic acid

PC3613-1G PC3613-5G PC3613-25G
APOSPC3613-1GEA 40.8 EUR
APOSPC3613-1G APOSPC3613-5G APOSPC3613-25G
2-Fluoro-3-methyl-5-pyridineboronic acid
2-Fluoro-3-methyl-5-pyridineboronic acid
Formula: C₆H₇BFNO₂
Peso molecolare: 154,94 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD03411562
CAS Number: 904326-92-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR