Stampa…

3-Fluoro-5-nitro-1-trifluoromethylbenzene 95%

Fornitore: Apollo Scientific

PC3601-10G PC3601-1G PC3601-5G
APOSPC3601-10GEA 219 EUR
APOSPC3601-10G APOSPC3601-1G APOSPC3601-5G
3-Fluoro-5-nitro-1-trifluoromethylbenzene 95%
3-Fluoro-5-nitro-1-trifluoromethylbenzene
Formula: C7H3F4NO2
Peso molecolare: 209,0978 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD07779168
CAS Number: 454-73-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR