Stampa…

3-Methyl-5-(pentafluorothio)benzyl bromide 98%

Fornitore: Apollo Scientific

Synonyms: 3-Methyl-5-(pentafluorosulphur)benzyl bromide

PC303071-1G PC303071-5G
APOSPC303071-1GEA 350 EUR
APOSPC303071-1G APOSPC303071-5G
3-Methyl-5-(pentafluorothio)benzyl bromide 98%
3-Methyl-5-(pentafluorothio)benzyl bromide
Formula: C₈H₈BrF₅S
Peso molecolare: 311,11 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16652481
CAS Number: 1240257-48-0
UN: 3261
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR