Stampa…

3,5-Difluoro-2-methoxybenzyl bromide 98%

Fornitore: Apollo Scientific
Pericolo

Synonyms: 1-(Bromomethyl)-3,5-difluoro-2-methoxybenzene

PC303053-10G PC303053-5G PC303053-1G
APOSPC303053-10GEA 184 EUR
APOSPC303053-10G APOSPC303053-5G APOSPC303053-1G
3,5-Difluoro-2-methoxybenzyl bromide 98%
3,5-Difluoro-2-methoxybenzyl bromide
Formula: C₈H₇BrF₂O
Peso molecolare: 237,04 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06660240
CAS Number: 886500-63-6
UN: 3265
ADR: 8,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR