2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile 97%
Fornitore: Apollo Scientific
PC303035-10G
PC303035-5G
PC303035-1G
APOSPC303035-10GEA
351
EUR
APOSPC303035-10G
APOSPC303035-5G
APOSPC303035-1G
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile 97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile
Formula:
C₉H₅F₄NO Peso molecolare: 219,14 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD22201044 CAS Number: 1373920-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...