Stampa…

3-Ethoxy-2,6-difluorobenzyl alcohol 97%

Fornitore: Apollo Scientific

Synonyms: (3-Ethoxy-2,6-difluorophenyl)methanol

PC302839-10G PC302839-5G PC302839-1G
APOSPC302839-10GEA 282 EUR
APOSPC302839-10G APOSPC302839-5G APOSPC302839-1G
3-Ethoxy-2,6-difluorobenzyl alcohol 97%
3-Ethoxy-2,6-difluorobenzyl alcohol
Formula: C₉H₁₀F₂O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11519370
CAS Number: 1092461-28-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR