Stampa…

5-Chloro-2-(trifluoromethoxy)phenol 97%

Fornitore: Apollo Scientific

PC302760-1G PC302760-5G
APOSPC302760-1GEA 170 EUR
APOSPC302760-1G APOSPC302760-5G
5-Chloro-2-(trifluoromethoxy)phenol 97%
5-Chloro-2-(trifluoromethoxy)phenol
Formula: C₇H₄ClF₃O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11519363
CAS Number: 1092461-24-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR