(4-Ethoxy-3,5-difluorophenyl)acetic acid 97%
Fornitore: Apollo Scientific
PC302750-1G
PC302750-5G
PC302750-10G
APOSPC302750-1GEA
159
EUR
APOSPC302750-1G
APOSPC302750-5G
APOSPC302750-10G
(4-Ethoxy-3,5-difluorophenyl)acetic acid 97%
(4-Ethoxy-3,5-difluorophenyl)acetic acid
Formula:
C₁₀H₁₀F₂O₃ Peso molecolare: 216,18 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09258681 CAS Number: 1017779-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...