3-Chloro-5-(trifluoromethyl)benzyl bromide
Fornitore: Apollo Scientific
|
Pericolo
|
PC302533-1G
PC302533-5G
PC302533-500MG
APOSPC302533-1GEA
50.9
EUR
APOSPC302533-1G
APOSPC302533-5G
APOSPC302533-500MG
3-Chloro-5-(trifluoromethyl)benzyl bromide
3-Chloro-5-(trifluoromethyl)benzyl bromide
Formula:
C₈H₅BrClF₃ Peso molecolare: 273.48 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04115849 CAS Number: 886496-91-9 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...