3-Chloro-4-(trifluoromethyl)cinnamic acid 97%
Fornitore: Apollo Scientific
PC302047-1G
PC302047-5G
APOSPC302047-1GEA
104
EUR
APOSPC302047-1G
APOSPC302047-5G
3-Chloro-4-(trifluoromethyl)cinnamic acid 97%
3-Chloro-4-(trifluoromethyl)cinnamic acid
Formula:
C₁₀H₆ClF₃O₂ Peso molecolare: 250,60 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11519328 CAS Number: 1092460-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...